Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146307.1 | NP_001139779.1 | 15 | 1766 | |
NM_024412.3 | NP_077723.3 | 16 | 1770 | ||
GenBank mRNA | AF124848.1 | - | 15 | 1863 | |
AY373832.1 | - | 15 | 1721 | ||
BC037077.1 | - | 16 | 1774 | 5042736 |