Gene Transcripts

Gene Symbol
Txnl4a
Entrez Gene ID
27366
Gene Name
thioredoxin-like 4A
Gene Aliases
D18Wsu98e, Dim1, ENSMUSG00000057130, Txnl4, U5-15kD, U5-15kDa
Chromosome Location
Chr. 18: 80206794 - 80225851 on Build GRCm38
UniGene ID
Mm.172411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025299.3 NP_079575.1 4 1176
GenBank mRNA AK004039.1 - 4 1133
AK148287.1 - 4 1159
AK166497.1 - 4 1152

View Full Product Details