Gene Transcripts

Gene Symbol
Hsd17b14
Entrez Gene ID
66065
Gene Name
hydroxysteroid (17-beta) dehydrogenase 14
Gene Aliases
0610039E24Rik, Dhrs10, retSDR3
Chromosome Location
Chr. 7: 45554855 - 45567321 on Build GRCm38
UniGene ID
Mm.285710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025330.3 NP_079606.3 8 612
GenBank mRNA AK002831.1 - 8 565
AK010556.1 - 4 277
BC064042.1 - 4 395 1332242
BC099530.1 - 4 281

View Full Product Details