Gene Transcripts

Gene Symbol
Sarnp
Entrez Gene ID
66118
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
1110005A23Rik, Cip29
Chromosome Location
Chr. 10: 128821766 - 128877638 on Build GRCm38
UniGene ID
Mm.351579
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025364.2 NP_079640.1 2 290
GenBank mRNA AK003453.1 - 2 142
AK018773.1 - 2 153
AK088266.1 - 2 139
AK146248.1 - 2 168
AK212934.1 - 3 174
AW824990.1 - 2 297
BC027510.1 - 2 129 1382354
BC094393.1 - 2 129
BC157909.1 - 2 154 9088111
BC158023.1 - 2 154 9007329
BC172063.1 - 2 154 9054349

View Full Product Details