Gene Transcripts

Gene Symbol
Tmem14c
Entrez Gene ID
66154
Gene Name
transmembrane protein 14C
Gene Aliases
1110021D01Rik, AU020193, HSPC194
Chromosome Location
Chr. 13: 41016250 - 41022582 on Build GRCm38
UniGene ID
Mm.30005
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025387.3 NP_079663.1 6 897
GenBank mRNA AK003891.1 - 6 846
AK008495.1 - 5 787
AK008878.1 - 6 819
AK010609.1 - 6 857
AK012101.1 - 6 844
AK019395.1 - 6 847
AK050688.1 - 6 832
AK089021.1 - 5 785
AK201301.1 - 1 269
BC025854.1 - 6 798 5250659
BC055862.1 - 6 824 3492090
BC069899.1 - 5 1543

View Full Product Details