Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025399.3 | NP_079675.1 | 5 | 580 | |
GenBank mRNA | AK003991.1 | - | 5 | 587 | |
AK013174.1 | - | 5 | 536 | ||
AK179685.1 | - | 1 | 36 | ||
AK205876.1 | - | 1 | 36 | ||
BC025444.1 | - | 5 | 582 | 3986887 |