Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025526.4 | NP_079802.1 | 1, 2 | 141 | |
GenBank mRNA | AK009021.1 | - | 1, 2 | 142 | |
AK009421.1 | - | 1, 2 | 145 | ||
AK039834.1 | - | 1, 2 | 183 | ||
AK083247.1 | - | 1, 2 | 149 | ||
AK157864.1 | - | 1, 2 | 150 | ||
BC028800.1 | - | 1, 2 | 119 | ||
BC048409.1 | - | 1, 2 | 138 | 6484953 | |
BC096049.1 | - | 1, 2 | 139 | 3466271 |