Gene Transcripts

Gene Symbol
Adamtsl5
Entrez Gene ID
66548
Gene Name
ADAMTS-like 5
Gene Aliases
2010109H09Rik, Thsd6
Chromosome Location
Chr. 10: 80339793 - 80349036 on Build GRCm38
UniGene ID
Mm.268527
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK008355.1 - 8 918
AK008364.1 - 8 918
AK008440.1 - 8 917
AK029081.1 - 8 894
AK154947.1 - 5 543
BC058577.1 - 4 1529

View Full Product Details