Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025670.4 | NP_079946.2 | 5 | 589 | |
GenBank mRNA | AK005157.1 | - | 5 | 575 | |
AK017480.1 | - | 5 | 572 | ||
AK031705.1 | - | 5 | 591 | ||
AK035791.1 | - | 3 | 1884 | ||
AK051560.1 | - | 5 | 587 | ||
AK149939.1 | - | 5 | 585 | ||
AK150219.1 | - | 5 | 534 | ||
AK166141.1 | - | 5 | 582 | ||
BC004063.1 | - | 5 | 588 | 3590511 |