Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025679.3 | NP_079955.1 | 4 | 572 | |
GenBank mRNA | AK014113.1 | - | 4 | 573 | |
AK014247.1 | - | 4 | 579 | ||
AK017452.1 | - | 4 | 572 | ||
AK017694.1 | - | 4 | 568 | ||
AK019264.1 | - | 4 | 558 | ||
AK033553.1 | - | 4 | 576 | ||
AK035590.1 | - | 4 | 586 | ||
AK038295.1 | - | 2 | 1759 | ||
BC004090.1 | - | 4 | 577 | 3593790 |