Gene Transcripts

Gene Symbol
Odf2l
Entrez Gene ID
52184
Gene Name
outer dense fiber of sperm tails 2-like
Gene Aliases
4733401D09Rik, 9630045K08Rik, D3Ertd250e
Chromosome Location
Chr. 3: 145118574 - 145153915 on Build GRCm38
UniGene ID
Mm.259105
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162538.1 NP_001156010.1 17 2298
NM_001162539.1 NP_001156011.1 15 2010
NM_025714.4 NP_079990.2 16 2005
GenBank mRNA AK014630.1 - 16 2007
AK220159.1 - 6 3482
BC020075.1 - 15 1979 3979963

View Full Product Details