Gene Transcripts

Gene Symbol
Thap2
Entrez Gene ID
66816
Gene Name
THAP domain containing, apoptosis associated protein 2
Gene Aliases
2900040O07Rik, 9030625G08Rik, AI450385, AI649097
Chromosome Location
Chr. 10: 115369966 - 115384435 on Build GRCm38
UniGene ID
Mm.32314
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025780.3 NP_080056.1 2 307
GenBank mRNA AK009162.1 - 2 286
AK018577.1 - 2 287
AK081303.1 - 2 308
BC027502.1 - 2 215 1515752

View Full Product Details