Gene Transcripts
- Gene Symbol
- Mfsd1
- Entrez Gene ID
- 66868
- Gene Name
- major facilitator superfamily domain containing 1
- Gene Aliases
- 1200003O06Rik
- Chromosome Location
- Chr. 3: 67582768 - 67604231 on Build GRCm38
- UniGene ID
- Mm.491153
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025813.3 |
NP_080089.1
|
4, 5 |
427 |
|
GenBank mRNA |
AK004586.1 |
- |
4, 5 |
428 |
|
AK032688.1 |
- |
4, 5 |
396 |
|
AK075859.1 |
- |
4, 5 |
457 |
|
AK135235.1 |
- |
4, 5 |
428 |
|
AK136108.1 |
- |
4, 5 |
419 |
|
AK152444.1 |
- |
4, 5 |
449 |
|
AK159554.1 |
- |
4, 5 |
455 |
|
AK218750.1 |
- |
2, 3 |
139 |
|
BC024891.1 |
- |
4, 5 |
409 |
3992247 |
View Full Product Details