Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025876.2 | NP_080152.1 | 2, 3 | 356 | |
GenBank mRNA | AK010081.1 | - | 2, 3 | 356 | |
AK010097.1 | - | 2, 3 | 356 | ||
AK088517.1 | - | 2, 3 | 365 | ||
AK136570.1 | - | 2, 3 | 369 | ||
BC025132.1 | - | 1, 2 | 321 | 3666341 |