Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025793.3 | NP_080069.2 | 6, 7 | 781 | |
GenBank mRNA | AK002352.1 | - | 6, 7 | 742 | |
AK013170.1 | - | 6, 7 | 731 | ||
AK146101.1 | - | 6, 7 | 782 | ||
AK151975.1 | - | 6, 7 | 743 | ||
AK168603.1 | - | 6, 7 | 734 | ||
AK170602.1 | - | 6, 7 | 728 | ||
BC004595.1 | - | 6, 7 | 725 | 3584839 |