Gene Transcripts

Gene Symbol
Nhlrc2
Entrez Gene ID
66866
Gene Name
NHL repeat containing 2
Gene Aliases
1200003G01Rik, AI835049, AV002846, AW496455
Chromosome Location
Chr. 19: 56548261 - 56603503 on Build GRCm38
UniGene ID
Mm.392509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025811.3 NP_080087.1 11 3680
GenBank mRNA AK004564.1 - 11 3327
AK033384.1 - 11 3681

View Full Product Details