Gene Transcripts

Gene Symbol
Ceacam14
Entrez Gene ID
67084
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 14
Gene Aliases
1600021E03Rik, 1600025E09Rik, AV033953
Chromosome Location
Chr. 7: 17812682 - 17815627 on Build GRCm38
UniGene ID
Mm.33709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025957.4 NP_080233.1 2 298
GenBank mRNA AK005390.1 - 2 299
AK005482.1 - 2 299
AK005533.1 - 2 299
BC099412.1 - 2 228 30787387

View Full Product Details