Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025993.2 | NP_080269.1 | 3 | 850 | |
GenBank mRNA | AK010995.1 | - | 3 | 851 | |
AK132010.1 | - | 3 | 852 | ||
AK139957.1 | - | 4 | 569 | ||
AK154531.1 | - | 3 | 851 | ||
BC026790.1 | - | 3 | 536 | 5148857 |