Gene Transcripts

Gene Symbol
Lyzl1
Entrez Gene ID
67328
Gene Name
lysozyme-like 1
Gene Aliases
1700038F02Rik, Lyc2, Lyzd1
Chromosome Location
Chr. 18: 4165832 - 4182236 on Build GRCm38
UniGene ID
Mm.158148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026092.3 NP_080368.1 5 686
GenBank mRNA AK006241.1 - 5 687
AK006630.1 - 5 687
BC048450.1 - 5 657 6774504

View Full Product Details