Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026239.2 | NP_080515.1 | 1 | 395 | |
GenBank mRNA | AK005180.1 | - | 1 | 230 | |
AK018535.1 | - | 1 | 232 | ||
AK042538.1 | - | 1 | 231 | ||
AK043881.1 | - | 1 | 230 | ||
BC042623.1 | - | 1 | 395 | 5369317 | |
BC050880.1 | - | 1 | 297 | 5690508 |