Gene Transcripts

Gene Symbol
Loh12cr1
Entrez Gene ID
67774
Gene Name
loss of heterozygosity, 12, chromosomal region 1 homolog (human)
Gene Aliases
5830457J20Rik, AW124643
Chromosome Location
Chr. 6: 134639512 - 134711184 on Build GRCm38
UniGene ID
Mm.41892
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170479.1 NP_001163950.1 2, 3 328
NM_026371.3 NP_080647.2 2, 3 470
GenBank mRNA AK007423.1 - 2, 3 327
AK018019.1 - 2, 3 386
AK089648.1 - 2, 3 334
AK151342.1 - 2, 3 336
AK169615.1 - 2, 3 342
BC023059.1 - 2, 3 429 5355560
DV652098.1 - 2, 3 220
DV658678.1 - 2, 3 330

View Full Product Details