Gene Transcripts

Gene Symbol
SGSH
Entrez Gene ID
6448
Gene Name
N-sulfoglucosamine sulfohydrolase
Gene Aliases
HSS, MPS3A, SFMD
Chromosome Location
Chr. 17: 80200668 - 80220400 on Build GRCh38
UniGene ID
Hs.31074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000199.3 NP_000190.1 8 1370
XM_005257582.3 XP_005257639.1 8 1479
XM_011525127.2 XP_011523429.1 9 1358
XM_017024952.1 XP_016880441.1 8 2215
GenBank mRNA AB209900.1 - 5 4165
AK095969.1 - 7 1224
AK222890.1 - 8 1303
AK291257.1 - 8 1303
BC047318.1 - 8 1295 5226903
U30894.1 - 8 1295

View Full Product Details