Gene Transcripts

Gene Symbol
Apopt1
Entrez Gene ID
68020
Gene Name
apoptogenic, mitochondrial 1
Gene Aliases
1700081D05Rik, 2810002N01Rik, Apop-1, Apop1
Chromosome Location
Chr. 12: 111713269 - 111755055 on Build GRCm38
UniGene ID
Mm.425762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163388.1 NP_001156860.1 4 446
NM_026511.2 NP_080787.1 4 446
GenBank mRNA AK002873.1 - 4 448
AK005403.1 - 4 434
AK010493.1 - 4 436
AK012648.1 - 4 444
AK029408.1 - 4 422
BC071262.1 - 4 446 6509633

View Full Product Details