Gene Transcripts

Gene Symbol
Yif1a
Entrez Gene ID
68090
Gene Name
Yip1 interacting factor homolog A (S. cerevisiae)
Gene Aliases
5330422J23Rik, 54TM, Yif1, Yif1p
Chromosome Location
Chr. 19: 5088538 - 5092879 on Build GRCm38
UniGene ID
Mm.44202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026553.4 NP_080829.1 2 318
GenBank mRNA AK011071.1 - 2 312
AK077329.1 - 2 304
AK160409.1 - 2 271
BC011117.1 - 2 260 4190535

View Full Product Details