Gene Transcripts
- Gene Symbol
- Ooep
- Entrez Gene ID
- 67968
- Gene Name
- oocyte expressed protein
- Gene Aliases
- 2410146L05Rik, Floped, Moep19, Sddr
- Chromosome Location
- Chr. 9: 78376903 - 78378588 on Build GRCm38
- UniGene ID
- Mm.26883
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB283026.1 |
- |
1, 2 |
175 |
|
AK010809.1 |
- |
1, 2 |
236 |
|
DQ238107.1 |
- |
1, 2 |
236 |
|
View Full Product Details