Gene Transcripts

Gene Symbol
Wls
Entrez Gene ID
68151
Gene Name
wntless homolog (Drosophila)
Gene Aliases
5031439A09Rik, AI173978, AI987742, Gpr177
Chromosome Location
Chr. 3: 159839677 - 159938670 on Build GRCm38
UniGene ID
Mm.6766
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026582.4 NP_080858.3 5 940
GenBank mRNA AK019886.1 - 6 979
AK029053.1 - 5 899
AK029104.1 - 5 897
AK029315.1 - 5 897
AK031444.1 - 5 897
AK044810.1 - 5 899
AK049842.1 - 5 897
AK077407.1 - 5 898
AK128912.1 - 5 896
AK159207.1 - 5 897
AK159977.1 - 5 899
AK190589.1 - 2 96
BC018381.1 - 4 524 4237619
BC075615.1 - 5 940 5703610

View Full Product Details