Gene Transcripts

Gene Symbol
Ms4a6d
Entrez Gene ID
68774
Gene Name
membrane-spanning 4-domains, subfamily A, member 6D
Gene Aliases
1110058E16Rik, AI447446, Ms4a11
Chromosome Location
Chr. 19: 11586606 - 11604804 on Build GRCm38
UniGene ID
Mm.290390
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026835.2 NP_081111.1 6 782
GenBank mRNA AF237911.1 - 6 744
AK004295.1 - 6 765
AK152859.1 - 6 763
AK156878.1 - 6 765
AY039037.1 - 6 744
BC018331.1 - 6 719 3964037
BC029738.1 - 6 726 4016611

View Full Product Details