Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026886.3 | NP_081162.1 | 3, 4 | 711 | |
GenBank mRNA | AK005085.1 | - | 3, 4 | 711 | |
AK053836.1 | - | 3, 4 | 443 | ||
AK165719.1 | - | 3, 4 | 429 | ||
BC056392.1 | - | 3, 4 | 701 | 6855144 | |
JN860221.1 | - | 3, 4 | 358 |