Gene Transcripts

Gene Symbol
Clec4b1
Entrez Gene ID
69810
Gene Name
C-type lectin domain family 4, member b1
Gene Aliases
1810046I24Rik, Aplra2, Clec4b, DCAR, DCARbeta, Dcar2
Chromosome Location
Chr. 6: 123049962 - 123071555 on Build GRCm38
UniGene ID
Mm.333374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027218.3 NP_081494.1 2, 3 251
GenBank mRNA AA914236.1 - 2, 3 251
AK007794.1 - 2, 3 208
AY230260.1 - 2, 3 215

View Full Product Details