Gene Transcripts

Gene Symbol
H1fnt
Entrez Gene ID
70069
Gene Name
H1 histone family, member N, testis-specific
Gene Aliases
1700026P10Rik, H1.7, H1t2, Hanp1, mH1t2
Chromosome Location
Chr. 15: 98255982 - 98257307 on Build GRCm38
UniGene ID
Mm.107718
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027304.2 NP_081580.2 1 363
GenBank mRNA AB016273.1 - 1 335
AK006402.1 - 1 102
AY496853.1 - 1 361
BC052363.1 - 1 349 6773647

View Full Product Details