Gene Transcripts

Gene Symbol
March5
Entrez Gene ID
69104
Gene Name
membrane-associated ring finger (C3HC4) 5
Gene Aliases
1810015H18Rik, 2310008I22Rik, 2700055A20Rik, 5730499H23Rik, E130202O05Rik, MARCH-V, MITOL, Marchv, Rnf153
Chromosome Location
Chr. 19: 37207545 - 37224457 on Build GRCm38
UniGene ID
Mm.175989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164336.1 NP_001157808.1 2 442
NM_001164337.1 NP_001157809.1 2 442
NM_027314.3 NP_081590.3 2 442
GenBank mRNA AK007511.1 - 2 278
AK009232.1 - 2 428
AK009364.1 - 2 441
AK021403.1 - 2 412
BC064752.1 - 2 166 30049608
BC107214.1 - 2 153
BC107215.1 - 2 154 40051628
BC132455.1 - 2 206 40130732
BC132457.1 - 2 205 40130734
BY070559.1 - 2 335

View Full Product Details