Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027460.2 | NP_081736.2 | 3 | 472 | |
GenBank mRNA | AK017628.1 | - | 3 | 384 | |
AK157423.1 | - | 3 | 473 | ||
AK187296.1 | - | 2 | 58 | ||
BC011293.1 | - | 3 | 447 | 3989313 | |
BQ950692.1 | - | 3 | 407 | ||
CK795124.1 | - | 3 | 481 |