Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028450.3 | NP_082726.2 | 5 | 444 | |
GenBank mRNA | AK014093.1 | - | 4 | 193 | |
AK017798.1 | - | 5 | 422 | ||
AK028490.1 | - | 4 | 360 | ||
AK036670.1 | - | 5 | 445 | ||
BC032154.1 | - | 5 | 615 | 5068339 | |
BY713087.1 | - | 4 | 191 |