Gene Transcripts

Gene Symbol
Iyd
Entrez Gene ID
70337
Gene Name
iodotyrosine deiodinase
Gene Aliases
0610009A07Rik, AI265638, Dehal1
Chromosome Location
Chr. 10: 3540279 - 3554877 on Build GRCm38
UniGene ID
Mm.24153
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027391.3 NP_081667.1 3 478
GenBank mRNA AK002363.1 - 3 479
BC023358.1 - 3 501 5064638

View Full Product Details