Gene Transcripts
- Gene Symbol
- Dhrs2
- Entrez Gene ID
- 71412
- Gene Name
- dehydrogenase/reductase member 2
- Gene Aliases
- 5430405K24Rik, Hep27
- Chromosome Location
- Chr. 14: 55124154 - 55241440 on Build GRCm38
- UniGene ID
- Mm.85484
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027790.2 |
NP_082066.2
|
11 |
1240 |
|
GenBank mRNA |
AK017274.1 |
- |
11 |
1242 |
|
View Full Product Details