Gene Transcripts

Gene Symbol
Csl
Entrez Gene ID
71832
Gene Name
citrate synthase like
Gene Aliases
1700007H16Rik
Chromosome Location
Chr. 10: 99757705 - 99759658 on Build GRCm38
UniGene ID
Mm.483162
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027945.3 NP_082221.2 1 786
GenBank mRNA AK005713.1 - 1 787
AK161295.1 - 1 592
BC050750.1 - 1 584 6775049

View Full Product Details