Gene Transcripts
- Gene Symbol
- Acbd6
- Entrez Gene ID
- 72482
- Gene Name
- acyl-Coenzyme A binding domain containing 6
- Gene Aliases
- 0610010G04Rik, 2610100E10Rik
- Chromosome Location
- Chr. 1: 155558098 - 155687316 on Build GRCm38
- UniGene ID
- Mm.288249
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145781.1 |
NP_001139253.1
|
1, 2 |
738 |
|
NM_026683.1 |
NP_080959.1
|
1, 2 |
738 |
|
NM_028250.3 |
NP_082526.2
|
1, 2 |
738 |
|
GenBank mRNA |
AK002470.1 |
- |
1, 2 |
739 |
|
AK011781.1 |
- |
1, 2 |
731 |
|
AK082937.1 |
- |
1, 2 |
739 |
|
AK165836.1 |
- |
1, 2 |
733 |
|
AK167959.1 |
- |
2, 3 |
679 |
|
AK209754.1 |
- |
1, 2 |
53 |
|
BC028537.1 |
- |
1, 2 |
483 |
|
BC061029.1 |
- |
1, 2 |
464 |
|
BC115580.1 |
- |
1, 2 |
330 |
40102241 |
BC115581.1 |
- |
1, 2 |
330 |
40102242 |
View Full Product Details