Gene Transcripts

Gene Symbol
Arhgap8
Entrez Gene ID
73167
Gene Name
Rho GTPase activating protein 8
Gene Aliases
3110043J09Rik
Chromosome Location
Chr. 15: 84720052 - 84772857 on Build GRCm38
UniGene ID
Mm.128411
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164627.1 NP_001158099.1 5, 6 412
NM_001164628.1 NP_001158100.1 4, 5 404
NM_001205334.1 NP_001192263.1 5, 6 432
NM_028455.4 NP_082731.2 4, 5 424
GenBank mRNA AK014171.1 - 4, 5 375
AK040142.1 - 4, 5 374
AK086816.1 - 4, 5 378
AK088242.1 - 4, 5 380
BC005563.1 - 3, 4 367 3965919
BC010306.1 - 4, 5 394
BY723460.1 - 4, 5 398

View Full Product Details