Gene Transcripts

Gene Symbol
Sema3d
Entrez Gene ID
108151
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3D
Gene Aliases
4631426B19Rik
Chromosome Location
Chr. 5: 12383166 - 12588943 on Build GRCm38
UniGene ID
Mm.89313
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028882.4 NP_083158.3 18 2479
GenBank mRNA AK028900.1 - 18 2261
AK031704.1 - 18 2091
AK052671.1 - 18 2480
AK163828.1 - 3 271
BC138131.1 - 18 2022 8861150

View Full Product Details