Gene Transcripts

Gene Symbol
Ifitm7
Entrez Gene ID
74482
Gene Name
interferon induced transmembrane protein 7
Gene Aliases
4933438K12Rik, Mil4
Chromosome Location
Chr. 16: 13981479 - 13986867 on Build GRCm38
UniGene ID
Mm.484026
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017110.1 - 1 1140
BC115786.1 - 1 436 40105118
BC115814.1 - 1 436 40105119

View Full Product Details