Gene Transcripts

Gene Symbol
Ddit4
Entrez Gene ID
74747
Gene Name
DNA-damage-inducible transcript 4
Gene Aliases
5830413E08Rik, AA415483, REDD1, Rtp801, dig2
Chromosome Location
Chr. 10: 59949675 - 59951770 on Build GRCm38
UniGene ID
Mm.21697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029083.2 NP_083359.1 3 1009
GenBank mRNA AK017926.1 - 3 1009
BC132645.1 - 3 1008 40130922
BC145370.1 - 3 1008 9053928

View Full Product Details