Gene Transcripts

Gene Symbol
KEL
Entrez Gene ID
3792
Gene Name
Kell blood group, metallo-endopeptidase
Gene Aliases
CD238, ECE3
Chromosome Location
Chr. 7: 142941114 - 142962416 on Build GRCh38
UniGene ID
Hs.368588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000420.2 NP_000411.1 4 547
XM_005249993.2 XP_005250050.1 4 398
GenBank mRNA AK314831.1 - 4 448
BC003135.1 - 4 547 2906155
BC050639.1 - 4 442 5742350
FN998902.1 - 3 337
HE572750.1 - 3 337
M64934.2 - 4 457

View Full Product Details