Gene Transcripts
- Gene Symbol
- Pih1d1
- Entrez Gene ID
- 68845
- Gene Name
- PIH1 domain containing 1
- Gene Aliases
- 1110061L23Rik, 4933413A04Rik, Nop17
- Chromosome Location
- Chr. 7: 45153835 - 45160065 on Build GRCm38
- UniGene ID
- Mm.181689
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278207.1 |
NP_001265136.1
|
2, 3 |
220 |
|
NM_001285904.1 |
NP_001272833.1
|
2, 3 |
212 |
|
NM_029406.4 |
NP_083682.1
|
2, 3 |
340 |
|
GenBank mRNA |
AK004341.1 |
- |
2, 3 |
221 |
|
AK016797.1 |
- |
2, 3 |
246 |
|
AK089003.1 |
- |
2, 3 |
289 |
|
AK146039.1 |
- |
2, 3 |
278 |
|
AK152695.1 |
- |
2, 3 |
209 |
|
AK177389.1 |
- |
1, 2 |
85 |
|
BC068254.1 |
- |
2, 3 |
229 |
30013096 |
View Full Product Details