Gene Transcripts
- Gene Symbol
- Vmn2r58
- Entrez Gene ID
- 628422
- Gene Name
- vomeronasal 2, receptor 58
- Gene Aliases
- EG628422
- Chromosome Location
- Chr. 7: 41836881 - 41872670 on Build GRCm38
- UniGene ID
- Mm.469902
- Species
- Mus musculus
- Gene Symbol
- Vmn2r-ps54
- Entrez Gene ID
- 75835
- Gene Name
- vomeronasal 2, receptor, pseudogene 54
- Gene Aliases
- 1700108E19Rik, 4930562N12Rik, Casr-rs5, ENSMUSG00000078527, Gm10948, Gprc2a-rs5
- Chromosome Location
- Chr. 7: 41668014 - 41727193 on Build GRCm38
- UniGene ID
- Mm.159597
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016194.1 |
- |
3 |
597 |
|
AK030272.1 |
- |
3 |
564 |
|
- Gene Symbol
- Vmn2r59
- Entrez Gene ID
- 628444
- Gene Name
- vomeronasal 2, receptor 59
- Gene Aliases
- EG628444
- Chromosome Location
- Chr. 7: 42011792 - 42058981 on Build GRCm38
- UniGene ID
- Mm.469901
- Species
- Mus musculus
- Gene Symbol
- Vmn2r61
- Entrez Gene ID
- 637873
- Gene Name
- vomeronasal 2, receptor 61
- Gene Aliases
- Casr-rs2, EG637873, Gprc2a-rs2
- Chromosome Location
- Chr. 7: 42260053 - 42300755 on Build GRCm38
- UniGene ID
- Mm.469899
- Species
- Mus musculus
- Gene Symbol
- Vmn2r90
- Entrez Gene ID
- 626942
- Gene Name
- vomeronasal 2, receptor 90
- Gene Aliases
- EG626942
- Chromosome Location
- Chr. 17: 17669137 - 17734167 on Build GRCm38
- UniGene ID
- Mm.404019
- Species
- Mus musculus
View Full Product Details