Gene Transcripts

Gene Symbol
Ankrd55
Entrez Gene ID
77318
Gene Name
ankyrin repeat domain 55
Gene Aliases
C030011J08Rik
Chromosome Location
Chr. 13: 112288451 - 112384002 on Build GRCm38
UniGene ID
Mm.159895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168403.1 NP_001161875.1 5 913
NM_001168404.1 NP_001161876.1 5 450
NM_001168405.1 NP_001161877.1 4 807
NM_029898.3 NP_084174.2 4 807
GenBank mRNA AK039553.1 - 4 808
AK045480.1 - 4 441
AK081197.1 - 5 451
AK138273.1 - 4 462
AK140417.1 - 5 777
AK155092.1 - 3 248
AK171119.1 - 3 248
BC152317.1 - 4 434 40088531
BC152536.1 - 4 434 40088530

View Full Product Details