Gene Transcripts

Gene Symbol
Ctsq
Entrez Gene ID
104002
Gene Name
cathepsin Q
Gene Aliases
1600010J02Rik
Chromosome Location
Chr. 13: 61035038 - 61040597 on Build GRCm38
UniGene ID
Mm.46088
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029636.3 NP_083912.2 4, 5 471
GenBank mRNA AF456461.1 - 4, 5 502
AY014776.1 - 4, 5 471
BC099415.1 - 4, 5 478 30786803

View Full Product Details