Gene Transcripts

Gene Symbol
Hdhd2
Entrez Gene ID
76987
Gene Name
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases
0610039H12Rik, 3110052N05Rik
Chromosome Location
Chr. 18: 76944115 - 76972171 on Build GRCm38
UniGene ID
Mm.236127
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039201.1 NP_001034290.1 5 568
NM_029826.2 NP_084102.1 5 529
GenBank mRNA AK002837.1 - 5 517
AK014208.1 - 5 523
AK137285.1 - 5 521
AK147794.1 - 550
AK212986.1 - 1 55
BC058177.1 - 5 503 5250739

View Full Product Details