Gene Transcripts

Gene Symbol
Ankrd55
Entrez Gene ID
77318
Gene Name
ankyrin repeat domain 55
Gene Aliases
C030011J08Rik
Chromosome Location
Chr. 13: 112288451 - 112384002 on Build GRCm38
UniGene ID
Mm.159895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168403.1 NP_001161875.1 7 1167
NM_001168404.1 NP_001161876.1 7 704
NM_029898.3 NP_084174.2 6 1061
GenBank mRNA AK045480.1 - 6 695
AK081197.1 - 7 705
AK138273.1 - 6 716
AK140417.1 - 7 1031
BC152317.1 - 6 688 40088531
BC152536.1 - 6 688 40088530

View Full Product Details