Gene Transcripts
- Gene Symbol
- Kcnip4
- Entrez Gene ID
- 80334
- Gene Name
- Kv channel interacting protein 4
- Gene Aliases
- AV032399, Calp, Calp250, KChIP4, KChIP4a
- Chromosome Location
- Chr. 5: 48389503 - 49525122 on Build GRCm38
- UniGene ID
- Mm.160172
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199242.1 |
NP_001186171.1
|
3, 4 |
488 |
|
NM_001199243.1 |
NP_001186172.1
|
2, 3 |
386 |
|
NM_001199244.1 |
NP_001186173.1
|
2, 3 |
529 |
|
NM_001199245.1 |
NP_001186174.1
|
2, 3 |
382 |
|
NM_030265.3 |
NP_084541.3
|
2, 3 |
314 |
|
GenBank mRNA |
AF305071.1 |
- |
2, 3 |
285 |
|
AF453243.1 |
- |
2, 3 |
222 |
|
AK039048.1 |
- |
2, 3 |
315 |
|
AK049662.1 |
- |
2, 3 |
266 |
|
AK133416.1 |
- |
2, 3 |
234 |
|
AK148685.1 |
- |
2, 3 |
531 |
|
AK165773.1 |
- |
2, 3 |
256 |
|
AY647240.1 |
- |
2, 3 |
265 |
|
BC051130.1 |
- |
3, 4 |
488 |
6532082 |
DQ148501.1 |
- |
3, 4 |
488 |
|
DQ148502.1 |
- |
1, 2 |
246 |
|
DQ148503.1 |
- |
2, 3 |
266 |
|
DQ148504.1 |
- |
2, 3 |
382 |
|
View Full Product Details