Gene Transcripts

Gene Symbol
Trim35
Entrez Gene ID
66854
Gene Name
tripartite motif-containing 35
Gene Aliases
0710005M05Rik, A430106H13Rik, AW046487, HLS5, Mair, NC8, mKIAA1098
Chromosome Location
Chr. 14: 66297025 - 66311424 on Build GRCm38
UniGene ID
Mm.43871
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029979.3 NP_084255.2 3 728
GenBank mRNA AB060155.1 - 3 672
AF145374.1 - 3 712
AK003000.1 - 3 687
AK032635.1 - 3 701
AK146541.1 - 2 179
AK163749.1 - 3 664
AK209912.1 - 1 77
AK217178.1 - 1 77
BC049105.1 - 3 678 6418046
BC053494.1 - 3 593
BC145783.1 - 3 722 40131086
BC145809.1 - 3 722 40131112

View Full Product Details